Transport case for Self Contained Breathing Apparatus
Lieferant: SCOTT SAFETY
2014810
SCTS2014810EA 0 EUR
SCTS2014810
Transport case for Self Contained Breathing Apparatus
Beutel Equipment Bags and Instrument Cases
This carrying case made from moulded HDPE is used to hold one breathing apparatus set complete with face mask and cylinder.
  • Closed cell foam gasket
  • PE strap with velcro
The case features an internal insert to hold the apparatus, internal securing strap, sealing gasket, carrying handle and double latch fasteners.