Pipettes, P-2, Aquisel
Lieferant: SELECTA
1780225
SELE1780225EA 0 EUR
SELE1780225
Pipettes, P-2, Aquisel
Pipetten Serologische Pipetten
Polystyrene pipettes with zero-closure system allows the pipette filling, preventing that the blood exceeds the zero point and also avoiding total or partial loss.
  • Pipette filling by pressure till the bottom of the tube
  • For erythrocyte sedimentation read E.S.R. Westergren method
  • Compatible with conventional tubes of 12×86
  • Automatic sealing makes it a disposable product
Verpackung: Cardboard package of 200 units. Supplied in 800 units boxes.