Analogue hotplates, COMBIPLAC
Lieferant: SELECTA
3000718
SELE3000718EA 1600 EUR
SELE3000718
Analogue hotplates, COMBIPLAC
Heizgeräte Heizplatten
COMBIPLAC heating plates are designed to work at temperatures adjustable to 400 °C. The regulation is electronic and the heating elements are placed in a circular position on the surface of the plate. Undeformable steel plate.
  • Non warping steel hotplate has heating elements embedded throughout the hotplate surface maintaining the heat distribution
  • Main case is made of AISI 304 stainless steel
  • Main power switch with “ON” indicator lamp
  • Electronic energy control from 0 to 100 %
  • An indicator lamp shows when the unit is hot
Connector at the back for an external digital thermometer ("Sensoterm" or "Electemp-TFT").