Low temperature Pt100 probe
Lieferant: SenseAnywhere
0096647494821
SENA0096647494821EA 0 EUR
SENA0096647494821
Low temperature Pt100 probe
Temperaturfühler
This smart probe featuring a digital SenseAnywhere Bus interface has been especially designed to be used in ultra-low environments for monitoring cryogenic applications down to -200 °C. The class A wire wound ceramic Pt100 sensor ensures accurate measurements over a wide temperature range. Due to the digital interface this probe with its own serial number and integrated electonics can be calibrated. Calibrated SAB sensors can be easily hot swapped offering a big advantage and cost savings for recalibration of sensors.
  • Accurate temperature measurement
  • Realtime: 24/7 data gives a complete audit trail
  • Alarms: notification via email or text when limits are exceeded
  • Secure data storage: access your data anywhere and anytime via the cloud
  • Seath made from stainless steel
Casing material connector: PA (UL 94 V-0)
Sheath: 100×3 mm
Seal pot: 6×30 mm
Connection size: Ø 11,5 mm (0,45")
4-pin SenseAnywhere Bus (SAB) interface