Naphthol AS-MX-phosphat

Lieferant: Serva
Achtung

30002.01
SERA30002.01EA 65.8 EUR
SERA30002.01
Naphthol AS-MX-phosphat
Naphthol AS-MX-phosphat
Formel: C₁₉H₁₈NO₅P
Lagertemperatur: Gefrierschrank
MDL: MFCD00042711
CAS-Nummer: 1596-56-1
EINECS: 216-480-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR