Siliconöl, 20 cSt DC 200

Lieferant: Serva

Synonyme: Dimethylpolysiloxane, PDMS, Dimethyl siloxane polymer (methyl silicones), Silicone oil, Dimethicone, Polydimethylsiloxane

35133.01
SERA35133.01EA 0 EUR
SERA35133.01
Siliconöl, 20 cSt DC 200
Siliconöl

Silicone DC 200 fluid

Dichte: 0,946…0.952 g/ml (25 °C)
MDL: MFCD00084411
CAS-Nummer: 63148-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR