Siliconöl, 100 cSt DC 200
Lieferant: Serva
Synonyme:
Dimethylpolysiloxane, PDMS, Dimethyl siloxane polymer (methyl silicones), Silicone oil, Dimethicone, Polydimethylsiloxane
35135.01
SERA35135.01EA
56.3
EUR
SERA35135.01
Siliconöl, 100 cSt DC 200
Siliconöl
Silicone DC 200 fluid
MDL:
MFCD00084411 CAS-Nummer: 63148-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...