Siliconöl, 350 cSt DC 200

Lieferant: Serva

Synonyme: Dimethylpolysiloxane, PDMS, Dimethyl siloxane polymer (methyl silicones), Silicone oil, Dimethicone, Polydimethylsiloxane

35136.01
SERA35136.01EA 61.9 EUR
SERA35136.01
Siliconöl, 350 cSt DC 200
Siliconöl

Silicone DC 200 fluid

Dichte: 0,966…0.972 g/ml (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00084411
CAS-Nummer: 63148-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR