5-Sulfosalicylsäure

Lieferant: Serva
Achtung

Synonyme: 2-Hydroxy-5-sulfobenzoesäure, 3-Carboxy-4-hydroxybenzolsulfonsäure

35706.01
SERA35706.01EA 63.3 EUR
SERA35706.01
5-Sulfosalicylsäure
5-Sulfosalicylsäure
Formel: C₇H₆O₆S
Molecular Weight: 218,19 g/mol
Schmelzpunkt: 120 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 97-05-2
EINECS: 202-555-6
UN: 2585
ADR: 8,III
Merck Index: 12,09138

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR