Xylencyanol FF

Lieferant: Serva
Achtung

Synonyme: C.I. 42135, Sodium hydrogen 4-[[4-(ethylamino)-m-tolyl][4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl]benzene-1,3-disulphonate, Sodium 4-((4-(ethylamino)-3-methylphenyl)(4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-sulphobenzenesulphonate, acid Blue 147

38505.01
SERA38505.01EA 0 EUR
SERA38505.01
Xylencyanol FF
Xylene Cyanole FF
Formel: C₂₅H₂₇N₂NaO₆S₂
Molecular Weight: 538,62 g/mol
Schmelzpunkt: 295 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040657
CAS-Nummer: 2650-17-1
EINECS: 220-167-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR