Syringes, Cavro
Lieferant: SGE Analytical Science
008140
SGEA008140EA 174.6 EUR
SGEA008140
Syringes, Cavro
Spritzen Spritzen für die Chromatographie
These cavro syringes are a type of instrument syringes used in chromatography.
  • Luer lock termination
  • Gas tight plunger with CAVRO XL mounting hole