Poloxamer 188, Sigma-Aldrich®

Lieferant: Merck

Synonyme: PEG-PPG-PEG, Kolliphor® P 188, Synperonic® F68, Lutrol® F68, Pluronic® F-68

15759-1KG
SIAL15759-1KGEA 107 EUR
SIAL15759-1KG
Poloxamer 188, Sigma-Aldrich®
Kolliphor® P 188 (Lutrol® F68, Pluronic® F-68, Synperonic® F68)

Poloxamer 188 is widely used as a thickening agent and gel binder.


The Poloxamer 188 grades are poloxamers from the extensive line of block copolymers.

Formel: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
Molecular Weight: 8400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082049
CAS-Nummer: 9003-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR