5,5-Dimethylbarbitursäure, Sigma-Aldrich®

Lieferant: Merck

39566-10G
SIAL39566-10GEA 47.9 EUR
SIAL39566-10G
5,5-Dimethylbarbitursäure, Sigma-Aldrich®
5,5-Dimethylbarbitursäure
Formel: C₆H₈N₂O₃
Molecular Weight: 156,14 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 24448-94-0
EINECS: 246-265-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR