Pluronic® F-68 Mn ~ 8,400, Sigma-Aldrich®

Lieferant: Merck

Synonyme: PEG-PPG-PEG, Kolliphor® P 188, Synperonic® F68, Lutrol® F68, Pluronic® F-68

M-Clarity™ Qualitätsstufe = MQ100

412325-250G
SIAL412325-250GEA 103 EUR
SIAL412325-250G
Pluronic® F-68 Mn ~ 8,400, Sigma-Aldrich®
Kolliphor® P 188 (Lutrol® F68, Pluronic® F-68, Synperonic® F68)

Pluronic® F-68 LF Pastille. Hard and soft surface cleaners, defoamers in coatings and water treatment. Lubricant in metal working, anti-foaming aid and extender for linear and cross-linked polyesters and polyurethanes.

Formel: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
Molecular Weight: 8400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082049
CAS-Nummer: 9003-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR