(Hydroxypropyl)methylcellulose (Hypromellose), M.W. ~86,000, 4,000 mPa.s (2% in H2O), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methocel K100, Hydroxypropyl methyl cellulose

M-Clarity™ Qualitätsstufe = MQ200

423203-100G
SIAL423203-100GEA 251 EUR
SIAL423203-100G
(Hydroxypropyl)methylcellulose (Hypromellose), M.W. ~86,000, 4,000 mPa.s (2% in H2O), Sigma-Aldrich®
Hydroxypropyl methyl cellulose (Hypromellose)
Formel: C₃H₈O₂·xCH₄O
MDL: MFCD00131360
CAS-Nummer: 9004-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR