4-Aminobenzamidoxim, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Aminobenzamide oxime, 4-Amino-N'-hydroxybenzimidamide

722189-1G
SIAL722189-1GEA 55.2 EUR
SIAL722189-1G
4-Aminobenzamidoxim, Sigma-Aldrich®
4-Aminobenzamidoxime
Formel: C₇H₉N₃O
Molecular Weight: 151,17 g/mol
MDL: MFCD08061151
CAS-Nummer: 277319-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR