4-Aminothiobenzamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Aminothiobenzoesäureamid

724904-1G
SIAL724904-1GEA 60.1 EUR
SIAL724904-1G
4-Aminothiobenzamid, Sigma-Aldrich®
4-Aminothiobenzamid
Formel: C₇H₈N₂S
Molecular Weight: 152,22 g/mol
MDL: MFCD00040927
CAS-Nummer: 4714-67-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR