DTBP (Dimethyl 3,3′-dithiopropionimidate dihydrochloride), Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Wang/Richard Reagenz, DTBP
M-Clarity™ Qualitätsstufe = MQ100
Cross-linking reagent
Reactant for:
Synthesis of glutathione-sensitive cross-linked polyethylenimine gene vector
Preparation of cyclodextrin-containing polymers designed for gene delivery
Crosslinking of chick oviduct progesterone-receptor subunits
Caution: The reagent is readily hydrolyzed at neutral pH. Avoid use of reducing agents during coupling reactions.
Other Notes: Note that the amidine linkage preserves original primary amine positive charge. The disulfide linkage is cleavable by mild reduction. Incorporates an eight atom linker.
Formel:
C₈H₁₆N₂O₂S₂·2HCl Molecular Weight: 309,28 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00043245 CAS-Nummer: 38285-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...