Penicillin : Streptomycin-Lösung in 0,9% NaCl, Hybri-Max™, Bioreagenzien, steril gefiltert
Lieferant: Merck
P7539-100ML
SIALP7539-100MLEA
87.7
EUR
SIALP7539-100ML
Penicillin : Streptomycin-Lösung in 0,9% NaCl, Hybri-Max™, Bioreagenzien, steril gefiltert
Penicillin : Streptomycin
This solution contains 10000 units penicillin and 10 mg/ml streptomycin in 0.9% NaCl.
Penicillin-Streptomycin is used to supplement cell culture media to control bacterial contamination and also suitable for hybridoma.
Lagertemperatur:
Gefrierschrank |
CAS-Nummer:
1406-05-9 (Penicillin) - 57-92-1 (Streptomycin) |
Spezifikation Testergebnisse
Appearance (Form) | Solution |
Appearance (Turbidity) | Clear |
Osmolality | 306 - 374 mOs/kg |
Potency - Penicillin | 10000 - 12000 U/ml |
Potency Streptomycin | 10 - 12 mg/ml |
Sterility | Pass |
Endotoxin Level at 10 ml/l | ≤0.5 EU/ml |
Cell Line Suspended Cell Line | Sp2 Mouse Myeloma |
% Cloning Performance | ≥80% |
Expiration Date Period 18 Months |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...