Puromycin Dihydrochlorid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
(S)-2-Amino-N-((2S,3S,4R,5R)-5-(6-(Dimethylamino)-9H-purin-9-yl)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)-3-(4-methoxyphenyl)propanamide dihydrochloride
M-Clarity™ Qualitätsstufe = MQ300
P9620-10ML
SIALP9620-10MLEA
534
EUR
SIALP9620-10ML
Puromycin Dihydrochlorid, Sigma-Aldrich®
Puromycin Dihydrochlorid
Formel:
C₂₂H₂₉N₇O₅·2HCl Molecular Weight: 544,44 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00012691 CAS-Nummer: 58-58-2 EINECS: 200-387-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...