Concrete inspection hammer
Lieferant: Industrial Physics
LD0500
SIMELD0500EA 759 EUR
SIMELD0500
Concrete inspection hammer
Beschichtungsprüfgeräte
Hammer used to test the quality of concrete, as well as other construction materials, for analysing resistance.
Standards: EN 12504-2, ASTM, C805, BS 1881.