Density meter
Lieferant: Industrial Physics
LD0560
SIMELD0560EA 4760 EUR
SIMELD0560
Density meter
Dichtemessgeräte
This densimeter is designed to perform automatic calculation of density and compensation of water temperature.
  • Able to measure floating sample in water
  • Volume of solid sample can be measured
  • Values automatically displayed after the elapsed time