Inspection mirror
Lieferant: Industrial Physics
LD3025
SIMELD3025EA 33.83 EUR
SIMELD3025
Inspection mirror
Spiegel
Ideal for visual inspection of hard-to-reach spots. Spots that are difficult to inspect are even more difficult to blast or coat.
  • With a simple inspection mirror a visual inspection behind stiffeners, under- or on top of I-beams or other awkward places is easy to perform
  • Made of clear glass that can be positioned by setting the double all angle ball joints for for 360° viewing
  • Ball joints are manufactured from spring steel to withstand stresses of constant repositioning and hold the mirror in any desired position