Coating conveyor ground tester
Lieferant: Industrial Physics
LD5900
SIMELD5900EA 118.97 EUR
SIMELD5900
Coating conveyor ground tester
Beschichtungsprüfgeräte
This coating conveyor ground tester is designed to detect whether items to be painted are sufficiently grounded and is generally used in electrostatic coating applications.