Analogue hygrometer
Lieferant: Industrial Physics
LI0055
SIMELI0055EA 1280 EUR
SIMELI0055
Analogue hygrometer
Hygrometer Hygrometer, analog
This high precision destructive hygrometer is designed for applications in concrete, sand cement, and other construction materials. The water present in the sample chemically reacts with calcium carbide produces pressure in the vessel, which is proportional to the residual moisture content.
  • Fast and reliable moisture determination
  • Insensitive to external influences
  • Standard splash- /dustproof
  • Direct reading of the CM% moisture
  • For printer documentation to protect against loss and follow-up costs
Lieferumfang: Supplied in carrying case with balance, pressure cylinder, calcium carbide and cleaning tools.