Probenbecher, Fibrin
Lieferant: Simport Scientific
B720-40
SIMPB720-40EA 56.6 EUR
SIMPB720-40
Probenbecher, Fibrin
Probenbehälter Probengefäße für Analyser
PE, translucent.
  • Good chemical resistance
  • Good temperature resistance
Precision molded for constant volume and uniform heat transfer.