KoolPlate™ Cooling Tray
Lieferant: Simport Scientific
M479-24
SIMPM479-24EA 533 EUR
SIMPM479-24
KoolPlate™ Cooling Tray
Kühlbehälter Tischkühler
The Simport KoolPlate™ is ideal for chilling and preparing paraffin blocks, for sectioning at the microtome or for performing other laboratory procedures.
  • For chilling and preparing paraffin blocks
  • Keeps paraffin blocks cold up to 4 hours
  • Tray made of high impact polystyrene for longevity
  • Insulator base made of styrofoam
The KoolPlate™ freezes overnight (16 hours) in a conventional freezer and the surface remains cold for up to 4 hours when used with the supplied styrofoam insulator base placed under the KoolPlate™. It will prolong cooling and prevent moisture accumulation. It is made of thick wall high impact polystyrene in order to ensure a sturdy construction that will last for years.
The KoolPlate™ is stackable, a great space-saving feature. Warning–Not for use with organic solvents such as toluene, xylene, acetone, etc.
Bitte beachten Not for use with organic solvents such as toluene, xylene, acetone, etc.