Chlorine water ∼0,4% Cl gesättigter Lösung USP Chlorine Test Solution
Lieferant: Spectrum Chemical
Gefahr
C-205-500ML
SPCMC-205-500MLEA 325 EUR
SPCMC-205-500ML
Chlorine water ∼0,4% Cl gesättigter Lösung USP Chlorine Test Solution
Chlorine water
Chlorine Water, Saturated Solution, Approx. 0.4 Percent Chlorine, U.S.P. Chlorine Test Solution is a Spectrum solution concentrate that allows the preparation of fresh, reliable standard volumetric solutions quickly and accurately.
Simply dilute the desired USP test solution concentrate to the specified volume (carefully following instructions provided with the package) and the resulting solution will have an exact Normality within 0.1%. Spectrum USP test solution concentrates are relied upon for fast, accurate results in life sciences and pharmaceutical quality control applications.
CAS-Nummer: 7782-50-5
UN: 1760
ADR: 8,III