Hydroxypropyl methylcellulose (Hypromellose), 3,550 mPa.s USP Substitution Type 2208

Lieferant: Spectrum Chemical

Synonyme: Methocel K100, Hydroxypropyl methyl cellulose

HY124-12KG
SPCMHY124-12KGEA 2690 EUR
SPCMHY124-12KG
Hydroxypropyl methylcellulose (Hypromellose), 3,550 mPa.s USP Substitution Type 2208
Hydroxypropyl methyl cellulose (Hypromellose)

Hypromellose, Substitution Type 2208, 3,550 mPa.s, USP.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₃H₈O₂·xCH₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD00131360
CAS-Nummer: 9004-65-3

Bestellen

Spezifikation Testergebnisse

Viscosity of a 2% Aqueous Solution @ 20D °C 2663 - 4970 mPa·s
Assay (Methoxy Groups) 19.0 - 24.0%
Assay (Hydroxypropoxy Groups) 4.0 - 12.0%
pH 5.0 - 8.0
Loss On Drying max. 5.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR