Soybean oil, epoxidised
Lieferant: Spectrum Chemical
S1179-500GM
SPCMS1179-500GMEA
174
EUR
SPCMS1179-500GM
Soybean oil, epoxidised
Soybean oil, epoxidised
Formel:
C₅₇H₉₈O₁₂ Molecular Weight: 975,399 g/mol |
MDL:
MFCD00163560 CAS-Nummer: 8013-07-8 |
Spezifikation Testergebnisse
Specific gravity at 25 °C | 0.988 - 0.998 |
Acid value | 0.50 |
Colour (APHA) | 125 |
Iodine Value | 2.00 |
Moisture | 0.10% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...