Cryopreservation media, HyClone™ HyCryo™
Lieferant: HyClone products (Cytiva)
SR30001.02
SR30001.02EA
369
EUR
SR30001.02
Cryopreservation media, HyClone™ HyCryo™
Zelleinfriermedien
Animal origin-free HyClone™ HyCryo cryopreservation media is designed for general use.
- 0,2 μm sterile filtered
- Serum free formulation
- Increased recovery of viable cells versus standard cryopreservation formulations