Ear punches used for notching or punching mouse or rat ears at weaning for easy identification.
Made of Stainless steel
Ergonomic handles for easy control
Non-clinical use only
Ear punches used for notching or punching mouse or rat ears at weaning for easy identification. Made of stainless steel, with ergonomic handles for easy control. For non-clinical use only.