Ear punch
Lieferant: STOELTING
56719
STOE56719EA 0 EUR
STOE56719
Ear punch
Biopsiestanzen
Ear punches used for notching or punching mouse or rat ears at weaning for easy identification.
  • Made of Stainless steel
  • Ergonomic handles for easy control
  • Non-clinical use only
Ear punches used for notching or punching mouse or rat ears at weaning for easy identification. Made of stainless steel, with ergonomic handles for easy control. For non-clinical use only.