1,3-Bis(2,6-diisopropylphenyl)imidazoliumchlorid ≥97%

Lieferant: Strem Chemicals, Inc.

Synonyme: 1,3-Bis(2,6-di-i-propylphenyl)imidazolium chloride, IPr.HCl

07-0590-2G
STRE07-0590-2GEA 533 EUR
STRE07-0590-2G
1,3-Bis(2,6-diisopropylphenyl)imidazoliumchlorid ≥97%
1,3-Bis(2,6-diisopropylphenyl)imidazoliumchlorid

NHC

Formel: C₂₇H₃₇ClN₂
Molecular Weight: 425,05 g/mol
MDL: MFCD02684545
CAS-Nummer: 250285-32-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR