Bis(cyclopentadienyl)titandichlorid ≥99%

Lieferant: Strem Chemicals, Inc.
Achtung

Synonyme: Titanocenedichloride, Bis(cyclopentadienyl)titanium(IV) dichloride, Titanocene dichloride

22-0200-25G
STRE22-0200-25GEA 180 EUR
STRE22-0200-25G
Bis(cyclopentadienyl)titandichlorid ≥99%
Bis(cyclopentadienyl)titandichlorid

Metallocenes, Derivatives & Cp Precursors

Formel: C₁₀H₁₀Cl₂Ti
Molecular Weight: 248,97 g/mol
Schmelzpunkt: 289…291 °C
MDL: MFCD00003723
CAS-Nummer: 1271-19-8
EINECS: 215-035-9
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR