1,1'-Bis(diisopropylphosphino)ferrocen ≥98%

Lieferant: Strem Chemicals, Inc.
Achtung

Synonyme: DiPPF, 1,1'-Bis(di-i-propylphosphino)ferrocene

26-0275-2G
STRE26-0275-2GEA 228 EUR
STRE26-0275-2G
1,1'-Bis(diisopropylphosphino)ferrocen ≥98%
1,1'-Bis(diisopropylphosphino)ferrocen

Phosphine

Formel: C₂₂H₂₈FeP₂
Molecular Weight: 410,25 g/mol
MDL: MFCD02684559
CAS-Nummer: 97239-80-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR