Bis(1,5-cyclooctadien)nickel(0) ≥98%

Lieferant: Strem Chemicals, Inc.
Gefahr

Synonyme: Bis(1,5-cyclooctadiene)nickel(0) (Wax encapsulated) (ca. 0.05mmol/capsule), Ni(COD)2

28-0010-2G
STRE28-0010-2GEA 161 EUR
STRE28-0010-2G
Bis(1,5-cyclooctadien)nickel(0) ≥98%
Bis(1,5-cyclooctadien)nickel(0)
Formel: C₁₆H₂₄Ni
Molecular Weight: 275,06 g/mol
Schmelzpunkt: 60 °C (N2) (Zers.)
MDL: MFCD00058902
CAS-Nummer: 1295-35-8
EINECS: 215-072-0
UN: 1325
ADR: 4.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR