Tetrakis(triphenylphosphin)palladium(0) ≥99% (99.9+%-Pd)

Lieferant: Strem Chemicals, Inc.

Synonyme: Pd(PPh3)4

46-2150-5G 46-2150.0005
STRE46-2150-5GEA 208 EUR
STRE46-2150-5G ABCR46-2150.0005
Tetrakis(triphenylphosphin)palladium(0) ≥99% (99.9+%-Pd)
Tetrakis(triphenylphosphin)palladium(0)
Formel: C₇₂H₆₀P₄Pd
Molecular Weight: 1155,58 g/mol
Schmelzpunkt: 103…107 °C
Dichte: 1 g/cm³ (20 °C)
MDL: MFCD00010012
CAS-Nummer: 14221-01-3
EINECS: 238-086-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR