Temperature controller, accessory for hotplates and stirrers, Stuart® SCT2, Cole-Parmer
Lieferant: Cole Parmer
04807-90
STUA04807-90EA 440 EUR
STUA04807-90
Temperature controller, accessory for hotplates and stirrers, Stuart® SCT2, Cole-Parmer
Regler Temperaturregler
All hotplate models are compatible with the Stuart temperature controller SCT2. The SCT2 provides precise temperature control in aqueous or oily samples up to a maximum temperature of 200 °C.
  • Automatically recognized when connected
  • Accurate setting of liquid temperature
  • Built-in holder for support rod and probe holder
  • Removable temperature sensor
  • Bright, easy-to-read LED display
  • Compact and lightweight
Designed for use with STUART UC and US range magnetic hotplate stirrers.
A range of accessories is available to complete your equipment: Support rod; PTFE probe (for applications requiring chemical resistance) and a metal probe.
Lieferumfang: Supplied with stainless steel temperature probe.