TraceCERT®, Supelco® Cyanide IC Standard
Lieferant: Merck
90157-100ML
SUPL90157-100MLEA 58.2 EUR
SUPL90157-100ML
TraceCERT®, Supelco® Cyanide IC Standard
Standards Anionische Standards Cyanid-Standards
1000 mg/L cyanide in water, prepared with high purity K₂[Zn(CN)₄] and water.
Cyanide exists mainly in the complex form [Zn(CN)₄]²⁻ in the water (basic) solution.
Standards: Certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). NIST or BAM Traceable.