Iod ≥99,8% (durch titrimetrische Analyse), ReagentPlus®, Supelco®

Lieferant: Merck

I3380-50G
SUPLI3380-50GEA 46.1 EUR
SUPLI3380-50G
Iod ≥99,8% (durch titrimetrische Analyse), ReagentPlus®, Supelco®
Iod
Formel: I₂
Molecular Weight: 253,81 g/mol
MDL: MFCD00011355
CAS-Nummer: 7553-56-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR