Cerex® PPBA SPE Column
Lieferant: Tecan
30162758
TCAT30162758EA 0 EUR
TCAT30162758
Cerex® PPBA SPE Column
Chromatographiesäulen
A special phenyl boronic acid chemistry on polymer base resin for better capacity and solvent use. PBA utilizes an unique covalent interaction ideal for catecholamine analysis.