Cerex® HPSAX 96 Well Plate RCH
Lieferant: Tecan
412-0401RCH
TCAT412-0401RCHEA 0 EUR
TCAT412-0401RCH
Cerex® HPSAX 96 Well Plate RCH
Mikrotiterplatten Festphasenextraktionsplatten
Polymeric strong anionic exchange chemistry.
  • Ion-exchange
  • Liquid handler compatible
20 µm polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in fast and uniform sample prep. Ideal for acidic compounds.