SPE Column, NBE TRACE-N20
Lieferant: Tecan
611-0051R-NBE
TCAT611-0051R-NBEEA 0 EUR
TCAT611-0051R-NBE
SPE Column, NBE TRACE-N20
Chromatographiesäulen
An end-capped C18 silica sorbent ideal for reverse-phase retention of neutral and polar analytes.
  • Reversed-phase
  • Liquid handler compatible
A 15 µm particulate with endcapped C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Formulated for 25-OH-VitD/steroid assays.