Cerex® OFXQ SPE Column
Lieferant: Tecan
635-0101R
TCAT635-0101REA 0 EUR
TCAT635-0101R
Cerex® OFXQ SPE Column
Chromatographiesäulen
Proprietary blend of ion-exchange and water-wettable polymer targeted towards oral fluid collection devices.
  • Ion-exchange
  • Liquid handler compatible
A proprietary polymer resin blend of ion exchange chemistry providing unique selectivity. Especially designed for scientists working with oral fluid matrixes.