SPE Column, NBE OFQC
Lieferant: Tecan
637-0051R-NBE
TCAT637-0051R-NBEEA 0 EUR
TCAT637-0051R-NBE
SPE Column, NBE OFQC
Chromatographiesäulen
A proprietary polymer resin blend of ion exchange chemistry providing unique selectivity. Especially designed for scientists working with oral fluid matrixes.