Cerex® ETGS 96 Well Plate, RCH
Lieferant: Tecan
642-0501RCH
TCAT642-0501RCHEA 0 EUR
TCAT642-0501RCH
Cerex® ETGS 96 Well Plate, RCH
Mikrotiterplatten Festphasenextraktionsplatten
A 10 µm micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.
  • Cerex
  • Ion-exchange
  • Liquid handler compatible
Formulated for ETS/ETGS Testings.
Verpackung: 1 piece per box