Blood collection tubes, VENOSAFE™
Lieferant: TERUMO
VF-054SLI
TERUVF-054SLIEA 27.8 EUR
TERUVF-054SLI
Blood collection tubes, VENOSAFE™
Klinisches Diagnosezubehör Klinische Probenentnahme und Probenvorbereitung Entnahme und Vorbereitung von Blutproben
PET, glass clear.
VENOSAFE™ consists of a glass-clear PET tube with a patented gas barrier and a safety closure composed of a cap and an inner stopper part. The cap functions like a safety shield during opening and reclosing. The inner stopper part erfectly seals the tube and guarantees a smooth blood collection and sampling. Draw marks improve comfort during blood collection and lab efficiency by assuring correct blood/additive ratios.