Sharps disposal container, Terusafe®
Lieferant: TERUMO
XX-Q1510V2 XX-Q2010V2 XX-Q2510V2 XX-Q2000V2 XX-Q2010V3 XX-Q2000V3 XX-Q1500V2
TERUXX-Q1510V2EA 484 EUR
TERUXX-Q1510V2 TERUXX-Q2010V2 TERUXX-Q2510V2 TERUXX-Q2000V2 TERUXX-Q2010V3 TERUXX-Q2000V3 TERUXX-Q1500V2
Sharps disposal container, Terusafe®
Behälter Abfallbehälter für spitze/scharfe Abfälle
These Terusafe® boxes are equipped with a variety of apertures enabling different options of sharps disposal.
  • Reduce the risk of percutaneous injuries
  • Cannot be pierced or reached into
  • Secure closure prevents needle spill while transported