Cleaning swabs, Pillow-tip™ Coventry™
Lieferant: Texwipe
52121
TEXW52121EA 0 EUR
TEXW52121
Cleaning swabs, Pillow-tip™ Coventry™
Tupfer Reinigungstupfer
These Pillow-tip™ Coventry™ swabs consist of polyester knit fabric which is held in place using a shrink tube. They are low in nonvolatile residue, ionic and particulate contamination, while providing exceptional strength and solvent resistance.
  • Seamless design is lint-free
  • High-strength, adhesive-free swab, durable head construction
  • Excellent solvent compatibility
Engineered for high solvent retention with the cleanliness of ISO Class 4 processed polyester.
Ideal for cleaning high end disk drive components, micro mechanical cleaning, aerospace, pharmaceutical and biotechnical industry.
Bitte beachten *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904