Savant™ SpeedVac™ SPD130DLX Medium Capacity Concentrator for Combinatorial Chemistry Applications
Lieferant: THERMO SAVANT
SPD130DLX-230
THSASPD130DLX-230EA 11100 EUR
THSASPD130DLX-230
Savant™ SpeedVac™ SPD130DLX Medium Capacity Concentrator for Combinatorial Chemistry Applications
Konzentratoren
The Thermo Scientific™ Savant™ SpeedVac™ SPD130DLX vacuum concentrator is a medium-capacity, vacuum concentrator with a modular design. It is compatible with a wide variety of solvents and offers resistance to TFA, DMSO, and other aggressive solvents used in combinatorial chemistry applications.
  • Proven reliability: Over 50 years of experience with the Savant SpeedVac family
  • Consistent drying time: Heated glass lid reduces drying times and prevents condensation
  • User-friendly: Intuitive interface with three preset and three user-defined programs for efficient operation
  • Safety assurance: Soft-close lid locks securely; hands-free operation with lid stay mechanism and audible alarm
  • Data transfer: USB port for downloading live run data enhances transferability
Our advanced solvent solutions are meticulously formulated to cater to a wide range of laboratory and industrial applications, including the removal of ethyl acetate from samples for CAT assays, the extraction of drug metabolites from solid-phase extracts, and the elution of analytes for evapouration processes. Featuring high-purity options such as Acetone, Chloroform, Ethyl Acetate, Hexane, and Methylene Chloride, our solvents ensure optimal performance, minimal interference, and maximum recovery of target compounds. These solvents are designed to deliver reliable, efficient, and reproducible results, making them indispensable in pharmaceutical, analytical, and various other scientific endeavors.